Order Entry
Spain
ContactUsLinkComponent
IMAGEXPRESS MICRO 4: ENVIRONMENT CONTROL AND LED - TRANSMITTED LIGHT. FOR BRIGHTFIELD IMAGING WITH 4-60X MAGNIFICATION OBJECTIVES, 240 VAC 1 * 1 items
Catalog # MLDV5046549
Supplier:  MOLECULAR DEVICES
IMAGEXPRESS MICRO 4: ENVIRONMENT CONTROL AND LED - TRANSMITTED LIGHT. FOR BRIGHTFIELD IMAGING WITH 4-60X MAGNIFICATION OBJECTIVES, 240 VAC 1 * 1 items
Catalog # MLDV5046549
Supplier:  MOLECULAR DEVICES
Frequently Bought Together