Order Entry
Spain
ContactUsLinkComponent
Extreme precision therm., with saddle, -58+30:0,1°C, enclosed scale, cap. round, colourless, toluene filled, 550x9-10mm, total immer sion, richter-top finish with button, suitable for government calibration 1 * 1 items
Catalog # AMARL53150-TOL
Supplier:  Amarell
Extreme precision therm., with saddle, -58+30:0,1°C, enclosed scale, cap. round, colourless, toluene filled, 550x9-10mm, total immer sion, richter-top finish with button, suitable for government calibration 1 * 1 items
Catalog # AMARL53150-TOL
Supplier:  Amarell
Frequently Bought Together