Order Entry
Spain
ContactUsLinkComponent
Setting point therm., similar DIN 12785, -38+50:1C, milk glass scale, cap. prismatic, colourless, red special liquid, Richter-top finish without button, upper part 195x10mm, lower part 165x5mm, immersion 180mm, null
Catálogo # AMARM242024-FL/VC
Supplier:  Amarell
Setting point therm., similar DIN 12785, -38+50:1C, milk glass scale, cap. prismatic, colourless, red special liquid, Richter-top finish without button, upper part 195x10mm, lower part 165x5mm, immersion 180mm, null
Catálogo # AMARM242024-FL/VC
Supplier:  Amarell
Frequently Bought Together