Order Entry
Spain
ContactUsLinkComponent
Precision therm., low temp., DIN 12778, -200+30:1C, solid stem, blue reflecting, pentane compound, 350x6-7mm, with ring at top, sui table for government verification 1 * 1 items
Catalog # AMARL33094-PENT
Supplier:  Amarell
Precision therm., low temp., DIN 12778, -200+30:1C, solid stem, blue reflecting, pentane compound, 350x6-7mm, with ring at top, sui table for government verification 1 * 1 items
Catalog # AMARL33094-PENT
Supplier:  Amarell
Frequently Bought Together