Order Entry
Spain
ContactUsLinkComponent
Therm. with standard ground joint 10/30, solid stem, white backed, -10+150:1°C, built-in length 105mm, upper part 200x6-7mm, red spe cial liquid 1 * 1 items
Catalog # AMARD264100-FL
Supplier:  Amarell
Therm. with standard ground joint 10/30, solid stem, white backed, -10+150:1°C, built-in length 105mm, upper part 200x6-7mm, red spe cial liquid 1 * 1 items
Catalog # AMARD264100-FL
Supplier:  Amarell
Frequently Bought Together