Order Entry
Spain
ContactUsLinkComponent
Thermometer similar to BS 593, F400C/76, -5+400:1C, solid stem, white backed, gallium filled, 415mm long, 76mm immersion, durable p igment, suitable for government verification 1 * 1 items
Catalog # AMARA403408-GA
Supplier:  Amarell
Thermometer similar to BS 593, F400C/76, -5+400:1C, solid stem, white backed, gallium filled, 415mm long, 76mm immersion, durable p igment, suitable for government verification 1 * 1 items
Catalog # AMARA403408-GA
Supplier:  Amarell
Frequently Bought Together