Order Entry
Spain
ContactUsLinkComponent
Pocket saccharimeter, 0-7:0,1%mas., with therm. 0+30:1C with red or blue special liquid, with correction scale, reading below, ref. temp. 20C, 275mm long 1 * 1 items
Catalog # AMARH847470-FL
Supplier:  Amarell
Pocket saccharimeter, 0-7:0,1%mas., with therm. 0+30:1C with red or blue special liquid, with correction scale, reading below, ref. temp. 20C, 275mm long 1 * 1 items
Catalog # AMARH847470-FL
Supplier:  Amarell
Frequently Bought Together