Order Entry
Spain
ContactUsLinkComponent
Sweet wort hydrometer, 0-24:0,1%mas, with therm. 0+30:1C with red or blue special liquid, with correction, reading below, ref. temp . 20C, 500mm long 1 * 1 items
Catalog # AMARH847338-FL
Supplier:  Amarell
Sweet wort hydrometer, 0-24:0,1%mas, with therm. 0+30:1C with red or blue special liquid, with correction, reading below, ref. temp . 20C, 500mm long 1 * 1 items
Catalog # AMARH847338-FL
Supplier:  Amarell
Frequently Bought Together