Order Entry
Spain
ContactUsLinkComponent
Thermometer similar to BS 1704, E/75, 0+160:1C, solid stem, white backed, red special liquid, 75mm immersion, 300mm long, durable p igment 1 * 1 items
Catalog # AMARA402108-FL
Supplier:  Amarell
Thermometer similar to BS 1704, E/75, 0+160:1C, solid stem, white backed, red special liquid, 75mm immersion, 300mm long, durable p igment 1 * 1 items
Catalog # AMARA402108-FL
Supplier:  Amarell
Frequently Bought Together