Order Entry
Spain
ContactUsLinkComponent
Hydrometer for liquefied gas, suitable for overpressure of 14bar, with therm. 0+30:1°C with red or blue special liquid, 0,600-0,650: 0,001g/cm , ref. temp. 15°C, 360mm long 1 * 1 items
Catalog # AMARH838764-FL
Supplier:  Amarell
Hydrometer for liquefied gas, suitable for overpressure of 14bar, with therm. 0+30:1°C with red or blue special liquid, 0,600-0,650: 0,001g/cm , ref. temp. 15°C, 360mm long 1 * 1 items
Catalog # AMARH838764-FL
Supplier:  Amarell
Frequently Bought Together