Order Entry
Spain
ContactUsLinkComponent
HayeSep A, 80/100, preconditioned, 25.4 mm coil, 2.62 feet (0.8 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for large valve oven 1 * 1 items
Catalog # HEWLG3591-81226
Supplier:  Agilent
HayeSep A, 80/100, preconditioned, 25.4 mm coil, 2.62 feet (0.8 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for large valve oven 1 * 1 items
Catalog # HEWLG3591-81226
Supplier:  Agilent
Frequently Bought Together