Order Entry
Spain
ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human CD209. 96wells/kit, with removable strips. 3002190000 CN 0.1 0.045 kg Yes 365 N 20 T2 N/A 1 * 1 KIT
Catalog # BSBTEK1632
Supplier:  Boster Bio
undefined
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human CD209. 96wells/kit, with removable strips. 3002190000 CN 0.1 0.045 kg Yes 365 N 20 T2 N/A 1 * 1 KIT
Catalog # BSBTEK1632
Supplier:  Boster Bio