Order Entry
Spain
Orders LinkContactUsLinkComponent
3-Octanone

3-Octanone

Bottle, 25 g
Catalog # APOSOR6025-25G
Flammable
Irritant
Supplier:   Apollo Scientific
3-Octanone
Catalog # APOSOR6025-25G
Supplier:  Apollo Scientific
CAS Number:  106-68-3
3-Octanone

Bottle, 25 g

CAS Number:  106-68-3
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:106-68-3
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00009515
  • Molecular Weight:128.21 g/mol
  • Boiling Pt:167…168 °C
  • Density:0.822 g/cm³ (25 °C)
  • Flash Pt:46 °C
  • Storage Temperature:Ambient
  • UN:2271
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications