Safety spectacles, Solus™ 1000
Improved anti-fog coating; anti-UV (2C-1.2), anti-scratch, anti-fog, Blue/black, Amber PC
Catalog # 111-2375
Supplier: 3M
Jump to:
Product Details & Documents
3M™ Solus™ 1000 series safety glasses feature a strong polycarbonate lens in a slim, stylish frame. These glasses feature a Scotchgard™ Anti-fog/anti-scratch coating, and a soft temple and nose bridge.
- Stylish and modern slim frame with colour accented soft temple components
- Strong polycarbonate lens for impact resistance
- Scotchgard™ Anti-Fog coating offers superior anti-fog and anti-scratch properties, even after washing multiple times
- Lightweight with soft cushioning on nose bridge
- Suitable for automatic cleaning, disinfection, and sterilisation by autoclaving at 121 °C
- Inner padding on temples for increased comfort and stability
- Excellent protection against ultraviolet (UV) radiation
Optional: Adjustable headband and foam or TPE gasket.
Certifications: EN 166
Documents
Specifications
- Type:Improved anti-fog coating; anti-UV (2C-1.2), anti-scratch, anti-fog
- Colour:Blue/black
- Seq:03
- Lenses:Amber PC
Accessories
Product Information
- Catalog #AvailabilityPrice
- Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the faceFoam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the faceMMMASOLUSSRLog in for availability1 * 5 UN - 39,90 €
Recomendaciones
Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".
De lo contrario, recibirá recomendaciones genéricas.