Objectives, Panthera
CCIS® Plan achromatic Phase objective UC PL Ph 100×/1,25/S-Oil (WD=0,16 mm), Ph3, 0,16 mm
MOTI1101001706511
: MOTIC
- Magnification:Ph3 100x/1,25 (i)
- Description:CCIS® Plan achromatic Phase objective UC PL Ph 100×/1,25/S-Oil (WD=0,16 mm), Ph3
- Seq:12
- Working distance:0,16 mm