Order Entry
Spain
ContactUsLinkComponent
KLF4 - Human shRNA lentiviral particles (4 unique 29mer target-specific shRNA, 1 scramble control), 0.5 ml each, >10
Catalog # ORIGTL316853V
Supplier:  OriGene
KLF4 - Human shRNA lentiviral particles (4 unique 29mer target-specific shRNA, 1 scramble control), 0.5 ml each, >10
Catalog # ORIGTL316853V
Supplier:  OriGene
Frequently Bought Together