Agilent J and W HP-INNOWax GC column, 30 m, 0.25 mm, 0.25 µm, LTM column module 1 * 1 items
Catalog # HEWL19091N-133ILTM
Supplier: Agilent
Agilent J and W HP-INNOWax GC column, 30 m, 0.25 mm, 0.25 µm, LTM column module 1 * 1 items
Catalog # HEWL19091N-133ILTM
Supplier: Agilent
Frequently Bought Together
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR® Standard Micro Syringes
Supplier: VWR Collection
Snap caps for snap cap vials, N 18 and N 22
Supplier: MACHEREY-NAGEL
Safety canisters
Supplier: S.C.A.T.
Silica gel 60, irregular 60 - 200 µm for flash chromatography
Supplier: VWR Chemicals
HPLC columns, LiChrospher®
Supplier: Merck
Cellulose microcrystalline for TLC plates
Supplier: Merck
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Avantor® Ultrasphere®, HPLC Columns
Supplier: Avantor
Exhaust filters, V3.0
Supplier: S.C.A.T.
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
VWR®, Snap Cap Vials
Supplier: VWR Collection
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Certified screw top vials and closures
Supplier: Agilent
Certified headspace crimp top vials
Supplier: Agilent