Order Entry
Spain
ContactUsLinkComponent
Agilent J and W GC packed column in UltiMetal tubing, length 2.46 ft (0.75 m), 1/8 inch od, 2 mm id, Carbosieve G packing, mesh size 60/80, preconditioned 1 * 1 items
Catalog # HEWLG3591-81186
Supplier:  Agilent
Agilent J and W GC packed column in UltiMetal tubing, length 2.46 ft (0.75 m), 1/8 inch od, 2 mm id, Carbosieve G packing, mesh size 60/80, preconditioned 1 * 1 items
Catalog # HEWLG3591-81186
Supplier:  Agilent
Frequently Bought Together