Mezcla 4 componentes 20mg/l cada uno Sc Ge Rh Ir en HNO3 2% HF TR HCl T
Catalog # CMPLRM001287L1
Supplier: CUSTOM MADE CHEMICALS LAB
Mezcla 4 componentes 20mg/l cada uno Sc Ge Rh Ir en HNO3 2% HF TR HCl T
Catalog # CMPLRM001287L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Jerricans, narrow neck, HDPE, UN Approved
Supplier: LAMAPLAST
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF