Order Entry
Spain
ContactUsLinkComponent
Biotrate 50ml digital burette. Thread A 45, adapters for A 32, S 40 and A 38 included. 1 pcs of aspiration tube included. Liquid pathway autoclavable at 121C 1 bar. Test certificate included. 1 * 1 items
Catalog # BHITLH-723082
Supplier:  Sartorius
Biotrate 50ml digital burette. Thread A 45, adapters for A 32, S 40 and A 38 included. 1 pcs of aspiration tube included. Liquid pathway autoclavable at 121C 1 bar. Test certificate included. 1 * 1 items
Catalog # BHITLH-723082
Supplier:  Sartorius
Frequently Bought Together