Order Entry
Spain
Orders LinkContactUsLinkComponent
CF® Dye BCN
CF® Dye BCN
CF®650 BCN, membrane permeability: Permeant, 653/673 nm, 0,5 mg
Catalog # 96027.
Supplier:   Biotium
CF® Dye BCN
Catalog # 96027.
Supplier:  Biotium
CAS Number:  
CF® Dye BCN
CF® Dye BCN
CF®650 BCN, membrane permeability: Permeant, 653/673 nm, 0,5 mg
CAS Number:  
Price
592,00 €
Log in for availability
1 * 0,5 mg
Product Details & Documents
CF® Dyes with a BCN group react with azides via a copper-free bioorthogonal reaction to label target molecules for surface staining of live cells or cell extracts. Membrane-permeant CF® Dye BCN conjugates for intracellular labelling reactions are also available.
  • Fluorescent labelling of azide groups on target molecules via copper-free bioorthonal chemistry
  • Alternative to fluorescently labelled DIBO and DBCO
  • Suitable for surface or intracellular labelling of live cells or cell extracts
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labelling
  • Available in several CF® Dye options including membrane-permeant forms for intracellular labelling
CF® Dyes BCN (bicyclo[6.1.0]nonyne) derivatives are alternatives to fluorescently labelled DIBO and DBCO. They react with azides to form 1,2,3-triazole by copper-free bioorthogonal 1,3-dipolar Huisgen cycloaddition. Useful for fluorescent surface labelling of live cells or cell extracts, particularly when there are concerns about native protein function loss with copper. Membrane-permeant CF® Dye BCN options are also available for intracellular labelling reactions.
Available CF® Dye BCN derivatives: CF®405S, CF®405M, CF®488A, CF®568, CF®594, CF®640R, CF®647, CF®680, and CF®680R including membrane-permeant CF®440 BCN, CF®500 BCN, and CF®650 BCN. Biotium's next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Specifications
Specifications
  • Pk:
    0,5 mg
  • Description:
    CF®650 BCN, membrane permeability: Permeant
  • Ex/Em:
    653/673 nm