Order Entry
Spain
ContactUsLinkComponent
Microcentrifuge tubes 1,5 mL with screw cap and O-Ring Seal Polypropylene (PP). With conical bottom. 1 * 1.000 items
Catalog # RATI5616027
Supplier:  RATIOLAB
Microcentrifuge tubes 1,5 mL with screw cap and O-Ring Seal Polypropylene (PP). With conical bottom. 1 * 1.000 items
Catalog # RATI5616027
Supplier:  RATIOLAB
Frequently Bought Together