Order Entry
Spain
ContactUsLinkComponent
This DNP (dinitrophenol) tyramide conjugate is used for tyramide signal amplification (TSA)1,2,3, which is an enzyme-mediated dete ction method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic ac 1 * 0,5 mg
Catalog # BTIU96019
Proveedor:  Biotium
This DNP (dinitrophenol) tyramide conjugate is used for tyramide signal amplification (TSA)1,2,3, which is an enzyme-mediated dete ction method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic ac 1 * 0,5 mg
Catalog # BTIU96019
Proveedor:  Biotium
Frequently Bought Together