Order Entry
Spain
ContactUsLinkComponent
For quantitative detection of rat beta IG-H3 in cell culture supernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1571
Supplier:  Boster Bio
undefined
For quantitative detection of rat beta IG-H3 in cell culture supernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1571
Supplier:  Boster Bio