Order Entry
Spain
Orders LinkContactUsLinkComponent
Safety spectacles, Solus™ 1000
Safety spectacles, Solus™ 1000
Improved anti-fog coating; anti-UV ‎(2C-1.2), anti-scratch, anti-fog, Blue/black, Grey PC
Catalog # 111-2374
Supplier:   3M
Safety spectacles, Solus™ 1000
Catalog # 111-2374
Supplier:  3M
Safety spectacles, Solus™ 1000
Safety spectacles, Solus™ 1000
Improved anti-fog coating; anti-UV ‎(2C-1.2), anti-scratch, anti-fog, Blue/black, Grey PC
Price
23,60 €
Log in for availability
1 * 1 UN
Product Details & Documents
3M™ Solus™ 1000 series safety glasses feature a strong polycarbonate lens in a slim, stylish frame. These glasses feature a Scotchgard™ Anti-fog/anti-scratch coating, and a soft temple and nose bridge.
  • Stylish and modern slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • Scotchgard™ Anti-Fog coating offers superior anti-fog and anti-scratch properties, even after washing multiple times
  • Lightweight with soft cushioning on nose bridge
  • Suitable for automatic cleaning, disinfection, and sterilisation by autoclaving at 121 °C
  • Inner padding on temples for increased comfort and stability
  • Excellent protection against ultraviolet (UV) radiation
Optional: Adjustable headband and foam or TPE gasket.
Certifications: EN 166
Specifications
  • Type:
    Improved anti-fog coating; anti-UV ‎(2C-1.2), anti-scratch, anti-fog
  • Colour:
    Blue/black
  • Seq:
    02
  • Lenses:
    Grey PC
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the face
    Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the face
    MMMASOLUSSR
    Log in for availability
    1 * 5 UN - 39,90 €