γ-Cyclodextrine ≥99.0% (by HPLC)
100 g
TCIAC0869-100G
: TCI
: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
- :17465-86-0
- :C₄₈H₈₀O₄₀
- :MFCD00009595
- :1297.1248 g/mol
- :14,02718
- :241-482-4
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Specific rotation [a]20/D:+176.0 to +180.0 deg (C=1, H2O) (calcd.on anh.substance)
- Water:max. 12.0 %
- Pk:100 g



