Order Entry
Spain
ContactUsLinkComponent
Cyanine 647 with an aminooxy reactive group could be used to label a target molecule with an aldehyde or ketone group. Aminooxy grou p readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent. 1 * 1 mg
Catalog # 96008.
Supplier:  Biotium
Cyanine 647 with an aminooxy reactive group could be used to label a target molecule with an aldehyde or ketone group. Aminooxy grou p readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent. 1 * 1 mg
Catalog # 96008.
Supplier:  Biotium
Frequently Bought Together