Order Entry
Spain
Orders LinkContactUsLinkComponent
Glycerine, liquid

Glycerine, liquid

1 L
Catalog # ICNA0215119491
Supplier:   MP Biomedicals
Glycerine, liquid
Catalog # ICNA0215119491
Supplier:  MP Biomedicals
CAS Number:  56-81-5
Glycerine, liquid
Glycerine, liquid

1 L

CAS Number:  56-81-5
Price
71,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
Specifications

Specifications

  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.