Order Entry
Spain
Orders LinkContactUsLinkComponent
Glycerine, liquid

Glycerine, liquid

500 mL
Catalog # ICNA0215119490
Supplier:   MP Biomedicals
Glycerine, liquid
Catalog # ICNA0215119490
Supplier:  MP Biomedicals
CAS Number:  56-81-5
Glycerine, liquid
Glycerine, liquid

500 mL

CAS Number:  56-81-5
Price
55,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
Specifications

Specifications

  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.