3,3',5,5'-Tetrametilbencidina, liquid substrate
Product Details & Documents
TMB is a chromogenic substrate for Horse Radish Peroxidase (HRP). MP Biolomedicals TMB Solution is a convenient, non toxic and ready to use stable chromogen solution suitable for peroxidase and pseudo-peroxidase reactions in ELISA, it provides consistent performance and sensitivity to deliver reproducible results
- Ready-to-Use, convenient
- Reproducible Results
- Consistent Performance and Sensitivity
- Stabilized chromogen solution
- For ELISA procedures and peroxidase and pseudo-peroxidase reactions
- Stable up to 24 months
- Non toxic.
Metabolism of the TMB substrate by HRP creates a soluble bluish reaction product that may be quantified by measuring absorbance at 370 or 665 nm. Reaction can be stopped by addition of 1 volume of 1N hydrocloric or sulfurc acid to yield a yellow reaction product that can be measured by absorbance at 450 nm (within one hour).
Caution: Research Use Only (RUO).
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Storage Temperature:Nevera
Specifications
Specification Test Results
- Identity Test:Passes
- Functionality Test :Passes
- pH:3.1 ± 0.5
- Expiration Date:Month/Day/Year
Specifications
- Env:1 L
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



