Order Entry
Spain
ContactUsLinkComponent
SONOREX TECHNIK RM 210 UH, 40 KHZ, ULTRASONIC BATH, 400 V 3N 50/60 HZ, ID 750X650X500 MM 245 L, WELDED TANK 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 2X1000 WEFF., WITH OVERFLO 1 * 1 items
Catalog # BAND8270-GB
SONOREX TECHNIK RM 210 UH, 40 KHZ, ULTRASONIC BATH, 400 V 3N 50/60 HZ, ID 750X650X500 MM 245 L, WELDED TANK 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 2X1000 WEFF., WITH OVERFLO 1 * 1 items
Catalog # BAND8270-GB
Frequently Bought Together