Order Entry
Spain
Orders LinkContactUsLinkComponent
Process documentaion
Segosoft Miele edition, solution USB: process documentation software for data transfer using USB stick: Comfort software, installation instructions, USB data logger module with 1,4 m lead and interface seriel cable to connect the unit and the recorder (3 m), USB device
Connection: 1 unit
W×D×H: 109×90×31 mm, G 788 (inc CD), PG 8583, PG 8583 CD, PG 8593
  MIEL10019220
 :   MIELE
Process documentaion
  MIEL10019220
 :  MIELE
Process documentaion
Process documentaion
Segosoft Miele edition, solution USB: process documentation software for data transfer using USB stick: Comfort software, installation instructions, USB data logger module with 1,4 m lead and interface seriel cable to connect the unit and the recorder (3 m), USB device
Connection: 1 unit
W×D×H: 109×90×31 mm, G 788 (inc CD), PG 8583, PG 8583 CD, PG 8593
1 * 1 UN
  • Description:
    Segosoft Miele edition, solution USB: process documentation software for data transfer using USB stick: Comfort software, installation instructions, USB data logger module with 1,4 m lead and interface seriel cable to connect the unit and the recorder (3 m), USB device
    Connection: 1 unit
    W×D×H: 109×90×31 mm
  • Seq:
    05
  • For:
    G 788 (inc CD), PG 8583, PG 8583 CD, PG 8593