Order Entry
Spain
ContactUsLinkComponent
Conductivity Standard 60,000 (µS/cm) 25C 1 * 500 mL
Conductivity standards, KCl, referenced at 25 °C
Catalog # REGECSKC60M
Supplier:  REAGECON
CAS Number:  
Conductivity standards, KCl, referenced at 25 °C
Catalog # REGECSKC60M
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard (KCl)
  • Seq:
    35
  • Conductivity:
    60000 µS/cm

Specifications

About this item

These conductivity standards are aqueous and so avoid the problems of matrix errors and high temperature dependency of nonaqueous conductivity standards.

  • With an accuracy of ±1,0% (except 1,3 µS)
  • Temperature dependence data, lot no. and expiry date are printed on product labels

These solutions standardised at 25 °C are directly traceable to N.I.S.T standard reference materials and are tested using an ISO 17025 accredited method.

Frequently Bought Together