Order Entry
Spain
Orders LinkContactUsLinkComponent
Trypsin, Reference Standards

Trypsin, Reference Standards

Trypsin Recombinant Porcine, USP reference standard, 2 x 1 mL, Vial, 2
Catalog # 1700013.
Supplier:   USP
Trypsin, Reference Standards
Catalog # 1700013.
Supplier:  USP
CAS Number:  
Trypsin, Reference Standards

Trypsin Recombinant Porcine, USP reference standard, 2 x 1 mL, Vial, 2

CAS Number:  
Price
2 320,00 €
Log in for availability
1 * 2 UN
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    2
  • Standards:
    USP reference standard
  • Description:
    Trypsin Recombinant Porcine
  • Pk Info:
    2 x 1 mL
  • UN No.:
    1789
  • Cat. No.:
    1700013.
  • Description:
    Trypsin Recombinant Porcine
  • CAS No.:
    9002-07-7

Specifications

Recomendaciones

Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".

De lo contrario, recibirá recomendaciones genéricas.