Order Entry
Spain
ContactUsLinkComponent
Urine and Vomit Spillage Kit is a multi use kit containing 6 shakers of our NQ64 absorbent granules, 15 plastic scoops / scrapers, 15 aprons, 15 pairs of synthetic gloves, 15 disposal bags, and a tub of alcohol surface wipes 1 * 1 KIT
Catalog # GUESH8625
Supplier:  GUEST
Urine and Vomit Spillage Kit is a multi use kit containing 6 shakers of our NQ64 absorbent granules, 15 plastic scoops / scrapers, 15 aprons, 15 pairs of synthetic gloves, 15 disposal bags, and a tub of alcohol surface wipes 1 * 1 KIT
Catalog # GUESH8625
Supplier:  GUEST
Frequently Bought Together