Inlet liners
Spilt, with wool, universal, 6,3 mm, 4,0 mm, 78,5
REST21079
: Restek
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
- Ext.Ø:6,3 mm
- Ø int.:4,0 mm
- Description:Spilt, with wool, universal
- Length:78,5
- Seq:041
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



