Order Entry
Spain
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Splitless, Siltek, for trace samples >2 µl, 6,5 mm, 4,0, 78,5 mm
Catalog # REST20773-214
Supplier:   Restek
Inlet liners
Catalog # REST20773-214
Supplier:  Restek
Inlet liners
Inlet liners

Splitless, Siltek, for trace samples >2 µl, 6,5 mm, 4,0, 78,5 mm

Price
199,00 €
Log in for availability
1 * 5 UN
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,5 mm
  • Ø int.:
    4,0
  • Description:
    Splitless, Siltek, for trace samples >2 µl
  • Length:
    78,5 mm
  • Seq:
    017
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.