Order Entry
Spain
Orders LinkContactUsLinkComponent
Glycerine base solution USP Test Solution (TS)

Glycerine base solution USP Test Solution (TS)

Plastic bottle, 100 mL
Catalog # 85348.180
Corrosive
Supplier:   VWR Chemicals
Glycerine base solution USP Test Solution (TS)
Catalog # 85348.180
Supplier:  VWR Chemicals
CAS Number:  56-81-5
Glycerine base solution USP Test Solution (TS)
Glycerine base solution USP Test Solution (TS)

Plastic bottle, 100 mL

CAS Number:  56-81-5
Price
206,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Ready to use test solution (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
  • Merck Index:13,04497
  • Melting Pt:18.6 °C
  • Density:1.2102 g/cm³ (20 °C)
  • Flash Pt:120 °C
  • Storage Temperature:Ambient
  • EINECS:0
  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.