Order Entry
Spain
Orders LinkContactUsLinkComponent
Inserts
Inserts
E 143, insert 1/4, mesh tray, for various items, with 2 hinged handles, 5 mm mesh spacing, 5 mm frame all round, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 255×230×55 mm
Catalog # MIEL3830310
Supplier:   MIELE
Inserts
Catalog # MIEL3830310
Supplier:  MIELE
Inserts
Inserts
E 143, insert 1/4, mesh tray, for various items, with 2 hinged handles, 5 mm mesh spacing, 5 mm frame all round, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 255×230×55 mm
Price
211,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications
  • Description:
    E 143, insert 1/4, mesh tray, for various items, with 2 hinged handles, 5 mm mesh spacing, 5 mm frame all round
  • Seq:
    7
  • W×D×H:
    255×230×55 mm
  • For:
    PG 8535/36, PG 8582, PG 8582 CD, PG 8592