Silver Nitrate 0.1 mol/l (0.1N) volumetric solution 1 * 1.000 mL
Catalog # APPC181464.1211
Supplier: PanReac AppliChem
Silver Nitrate 0.1 mol/l (0.1N) volumetric solution 1 * 1.000 mL
Catalog # APPC181464.1211
Supplier: PanReac AppliChem
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Potassium hydrogen sulphate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Sulphuric acid 95-97%, EMSURE® ISO for analysis, Supelco®
Supplier: Merck
VWR® Media Supplements for Dehydrated Culture Media
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
Heptane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Sulphuric acid 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Filter funnels
Supplier: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
Magnetic stirrer heads, BOLA
Supplier: Bohlender
Sodium Thiosulfate 0.1 mol/l (0.1N) (Reag. Ph.Eur.) volumetric solution 1 * 1 L
Supplier: PanReac AppliChem