Perchloric Acid 0.1 mol/l (0.1N) in 1,4-dioxan (Reag. USP) volumetric solution 1 * 1.000 mL
APPC181047.1611
Perchloric Acid 0.1 mol/l (0.1N) in 1,4-dioxan (Reag. USP) volumetric solution 1 * 1.000 mL
APPC181047.1611
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
White spirit, 150…200 °C, TECHNICAL
: VWR Chemicals
Isoparaffine G analytical reagent
: PanReac AppliChem