VWR®, Stoppers, PE
Blue, 16 mm
Catalog # 212-0552
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
- Choice of colours
Specifications
- Ø top:16 mm
- Colour:Blue