Order Entry
Spain
ContactUsLinkComponent
Pocketmop in white microfiber, 18” width. for use with qpsl-18 adapter. suitable for use in controlled environments and support area s. compatible with strong disinfectants, launderable, autoclavable. packaged in packs of 4, 6 packs to a case. 1 * 4 items
Catalog # MICRPKMAW-18
Supplier:  MICRONOVA
Pocketmop in white microfiber, 18” width. for use with qpsl-18 adapter. suitable for use in controlled environments and support area s. compatible with strong disinfectants, launderable, autoclavable. packaged in packs of 4, 6 packs to a case. 1 * 4 items
Catalog # MICRPKMAW-18
Supplier:  MICRONOVA
Frequently Bought Together