Order Entry
Spain
ContactUsLinkComponent
ChemiDoc-It 2 515 Cam/Dkrm Kit: 515 camera: 4.1 mp (with fixed lens), 16 bit monochrome, digital camera with mount, with binning cap abilities, cooling to -57C from ambient. MOTORISED fixed 30mm, f/1.4 lens, close up diopter, 50x50mm Ethidium Bromi 1 * 1 items
Catalog # UVPA97-0693-04
undefined
ChemiDoc-It 2 515 Cam/Dkrm Kit: 515 camera: 4.1 mp (with fixed lens), 16 bit monochrome, digital camera with mount, with binning cap abilities, cooling to -57C from ambient. MOTORISED fixed 30mm, f/1.4 lens, close up diopter, 50x50mm Ethidium Bromi 1 * 1 items
Catalog # UVPA97-0693-04
Frequently Bought Together