Order Entry
Spain
ContactUsLinkComponent
Splitter kit, 75 to 300 mL/min, preparative, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0 .25 mm, 250 mm (5023-2263), tubing, solvent, 1.5 mm id, 3 mm od, 5 m (5062-2483), adaptor, PTFE, 1/4-28 female to 10- 1 * 1 items
Catalog # HEWL5023-2259
Supplier:  Agilent
Splitter kit, 75 to 300 mL/min, preparative, for Agilent G4260B evaporative light scattering detector. Includes capillary to ELSD, 0 .25 mm, 250 mm (5023-2263), tubing, solvent, 1.5 mm id, 3 mm od, 5 m (5062-2483), adaptor, PTFE, 1/4-28 female to 10- 1 * 1 items
Catalog # HEWL5023-2259
Supplier:  Agilent
Frequently Bought Together