Order Entry
Spain
ContactUsLinkComponent
Pharma-50 Tubing, Silicone, Dow Corning, Marked Tubing, 50ft, I.D. 0.125 x O.D. 0.250 x Wall Thickness 0.063 1 * 50 Feet
Catalog # DOWC3100430
Supplier:  DOW CORNING
Pharma-50 Tubing, Silicone, Dow Corning, Marked Tubing, 50ft, I.D. 0.125 x O.D. 0.250 x Wall Thickness 0.063 1 * 50 Feet
Catalog # DOWC3100430
Supplier:  DOW CORNING