Order Entry
Spain
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use

125 mL
Catalog # J63202.XE
Flammable
Health
Irritant
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
Catalog # J63202.XE
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use

125 mL

CAS Number:  54827-17-7
Price
199,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
  • ADR:3,II
Specifications

Specifications

  • Pk:
    125 mL

Specifications

Recomendaciones

Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".

De lo contrario, recibirá recomendaciones genéricas.