Order Entry
Spain
ContactUsLinkComponent
Oncostatin-M (209 a.a.) Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 209 amino ac ids and having a molecular mass of 23.9kDa. The Oncostatin-M (209 a.a.) is purified by proprietary chromatog 1 * 2 µG
Catalog # BIRBORB80096-2
Supplier:  Biorbyt
Oncostatin-M (209 a.a.) Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 209 amino ac ids and having a molecular mass of 23.9kDa. The Oncostatin-M (209 a.a.) is purified by proprietary chromatog 1 * 2 µG
Catalog # BIRBORB80096-2
Supplier:  Biorbyt