Order Entry
Spain
ContactUsLinkComponent
Endothelial Mitogen can be used for Human Mammalian Vascular Endothelial Cells at concentration of 0.1-0.3mg/ml of serum supplemente d media 0.01-0.1mg with heparin. Endothelial Mitogen can be used for Hybridoma Cell Cloning at concentration 1 * 100 mg
Catalog # BIRBORB81077-100
Supplier:  Biorbyt
Endothelial Mitogen can be used for Human Mammalian Vascular Endothelial Cells at concentration of 0.1-0.3mg/ml of serum supplemente d media 0.01-0.1mg with heparin. Endothelial Mitogen can be used for Hybridoma Cell Cloning at concentration 1 * 100 mg
Catalog # BIRBORB81077-100
Supplier:  Biorbyt