Order Entry
Spain
ContactUsLinkComponent
IL12 Rat Recombinant produced in Sf9 insect cells is glycosylated disulfide linked heterodimeric polypeptide containing 503 amino ac ids and having molecular weight of 70 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subuni 1 * 100 µG
Catalog # BIRBORB80439-100
Supplier:  Biorbyt
IL12 Rat Recombinant produced in Sf9 insect cells is glycosylated disulfide linked heterodimeric polypeptide containing 503 amino ac ids and having molecular weight of 70 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subuni 1 * 100 µG
Catalog # BIRBORB80439-100
Supplier:  Biorbyt